Thermo Scientific™

MDS Mobile Detection Systems

Catalog number: 4255009
Thermo Scientific™

MDS Mobile Detection Systems

Catalog number: 4255009
The Thermo Scientific™ MDS Mobile Detection System is a mobile radiation reconnaissance system designed to operate in a vehicle for use on the ground or in airspace. It is for use around or in airspace to discover/map the location of artificial radiation sources or survey large areas of contamination.
 
Catalog Number
4255009
Unit Size
Each
Description
Gamma Radiation Reconnaissance
Price (USD)
Full specifications
DescriptionGamma Radiation Reconnaissance
Height (English)13.8 in.
Height (Metric)35cm
Depth (English)15.7 in.
Depth (Metric)40 cm
Measurement RangesScintillation Detector: 1nSv/h to 20 µSv/h; Proportional Detector: 10nSv/h to 1 Sv/h
Radiation TypeGamma
Weight (English)33.1 lb.
Weight (Metric)15kg
Width (English)23.6 in.
Width (Metric)60cm
Item DescriptionGamma radiation reconnaissance
Relative Humidity5 to 95%, non-condensing
Temperature (English) Range32° to 122°F
Temperature (Metric) Range0° to 50°C
Threshold Limit Value50keV
TypeMobile Detection System
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Mobile Radiation Large Area Detection System
Price (USD)
Full specifications
DescriptionMobile Radiation Large Area Detection System
Detector TypeScintillation Detector
Height (English)13.8 in.
Height (Metric)35cm
Depth (English)15.7 in.
Depth (Metric)40 cm
Measurement RangesScintillation Detector: 1nSv/h to 20 µSv/h; Proportional Detector: 10nSv/h to 1 Sv/h
Radiation TypeDependent on probe used
Sensitivity2μSv/h (0.2μR/h) increase above background
Weight (English)33.1 lb.
Weight (Metric)15kg
Width (English)23.6 in.
Width (Metric)60cm
Item DescriptionMobile radiation detection system
Relative Humidity5 to 95, non-condensing %
Temperature (English) Range32° to 122°F
Temperature (Metric) Range0° to 50°C
Threshold Limit Value50keV
TypeMobile Detection System
Unit SizeEach
Showing 2 of 2
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
4255009Full specifications
EachGamma Radiation ReconnaissanceRequest A Quote
DescriptionGamma Radiation Reconnaissance
Height (English)13.8 in.
Height (Metric)35cm
Depth (English)15.7 in.
Depth (Metric)40 cm
Measurement RangesScintillation Detector: 1nSv/h to 20 µSv/h; Proportional Detector: 10nSv/h to 1 Sv/h
Radiation TypeGamma
Weight (English)33.1 lb.
Weight (Metric)15kg
Width (English)23.6 in.
Width (Metric)60cm
Item DescriptionGamma radiation reconnaissance
Relative Humidity5 to 95%, non-condensing
Temperature (English) Range32° to 122°F
Temperature (Metric) Range0° to 50°C
Threshold Limit Value50keV
TypeMobile Detection System
Unit SizeEach
4255004Full specifications
EachMobile Radiation Large Area Detection SystemRequest A Quote
DescriptionMobile Radiation Large Area Detection System
Detector TypeScintillation Detector
Height (English)13.8 in.
Height (Metric)35cm
Depth (English)15.7 in.
Depth (Metric)40 cm
Measurement RangesScintillation Detector: 1nSv/h to 20 µSv/h; Proportional Detector: 10nSv/h to 1 Sv/h
Radiation TypeDependent on probe used
Sensitivity2μSv/h (0.2μR/h) increase above background
Weight (English)33.1 lb.
Weight (Metric)15kg
Width (English)23.6 in.
Width (Metric)60cm
Item DescriptionMobile radiation detection system
Relative Humidity5 to 95, non-condensing %
Temperature (English) Range32° to 122°F
Temperature (Metric) Range0° to 50°C
Threshold Limit Value50keV
TypeMobile Detection System
Unit SizeEach
Showing 2 of 2
  • Rugged, easily relocated military specification system with user friendly features
  • Allows for a wide range of users without compromising accuracy and results
  • Remote viewing and/or control of mission from command station via wireless transmission of data
  • Fully integrated ArcView GIS for simultaneous hazard modeling and verification with real time data
  • Fleet management capabilities allows for use of many detectors and/or personnel and asset location
  • Lightweight, modular design allows for easy relocation between vehicles
  • No hard wiring into vehicles is required
  • Real time data feed into any version
  • ESRI ArcView software is standard
  • Optional neutron and contamination probes to detect and measure all types of radiation
  • NBR technology for instant threat assessment of radiological incidents
  • Data transmission by radio modem or GSM wireless devices
Ordering Alerts:
  • Optional Neutron Detector is He3 detector filled to 10 atmospheres
Includes:
  • Zargus military grade carrying case, NATO approved
Recommended for:
  • Tactical hazmat operations for civilian and military responders (CST's, police, fire, hazmat and radiological response teams)
  • Surveillance and intelligence gathering
  • Geo spatial intelligence analysis
  • Special event monitoring
  • Aerial surveys of contaminated areas

Figures

Frequently asked questions (FAQs)

Citations & References