2-Amino-4'-methoxyacetophenone hydrochloride, 97%
2-Amino-4'-methoxyacetophenone hydrochloride, 97%
2-Amino-4'-methoxyacetophenone hydrochloride, 97%
Acros Organics

2-Amino-4'-methoxyacetophenone hydrochloride, 97%

CAS: 3883-94-1 | C9H11NO2·ClH | 201.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 425710010
also known as 42571
HSN 29225090
Price (INR)
9,044.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
9,044.00
Each
Chemical Identifiers
CAS3883-94-1
IUPAC Name2-amino-1-(4-methoxyphenyl)ethan-1-one hydrochloridyl
Molecular FormulaC9H11ClNO2
InChI KeyZSUGWZZUIKUPMO-UHFFFAOYSA-N
SMILES[Cl].COC1=CC=C(C=C1)C(=O)CN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to tan
Infrared spectrumConforms
HPLC>=96.0 %
Titration Argentometric>=96.0 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only