BOC-DL-Valine, 98%
BOC-DL-Valine, 98%
BOC-DL-Valine, 98%
Thermo Scientific Chemicals

BOC-DL-Valine, 98%

CAS: 54895-12-4 | C10H19NO4 | 217.27 g/mol
Have Questions?
Catalog NumberQuantityPackaging
42578050050 gGlass bottle
Catalog number 425780500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS54895-12-4
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}-3-methylbutanoic acid
Molecular FormulaC10H19NO4
InChI KeySZXBQTSZISFIAO-UHFFFAOYSA-N
SMILESCC(C)C(NC(=O)OC(C)(C)C)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
NMRconsistent with structure
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only