Acetic acid, ammonium salt-d<sub>7</sub>, 99 atom % D, Thermo Scientific Chemicals
Acetic acid, ammonium salt-d<sub>7</sub>, 99 atom % D, Thermo Scientific Chemicals
Acetic acid, ammonium salt-d<sub>7</sub>, 99 atom % D, Thermo Scientific Chemicals
Acros Organics

Acetic acid, ammonium salt-d7, 99 atom % D, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 425860010
also known as 42586
HSN 29152990
Price (INR)
7,237.00
Each
Quantity:
1 g
Request bulk or custom format
Price (INR)
7,237.00
Each
Chemical Identifiers
CAS194787-05-8
IUPAC Nameammonium acetate
Molecular FormulaC2H7NO2
InChI KeyUSFZMSVCRYTOJT-UHFFFAOYSA-N
SMILES[NH4+].CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
D-Enrichment>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only