Nunc™ PETG Roller Bottles
Thermo Scientific™

Nunc™ PETG Roller Bottles

Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCell Growth AreaBottle SurfacePackaging
4260-224200 cm²Pleated22/Cs.
1060-051050 cm²Smooth5/Sleeve, 20/Cs.
1060-201050 cm²Smooth20/Cs.
1060-851050 cm²Smooth5/Sleeve, 20/Cs.
1760-201700 cm²Pleated20/Cs.
1860-221800 cm²Smooth22/Cs.
2160-202100 cm²Pleated20/Cs.
2160-052100 cm²Pleated5/Sleeve, 20/Cs.
Catalog number 4260-22
Price (MXN)
-
Cell Growth Area:
4200 cm²
Bottle Surface:
Pleated
Packaging:
22/Cs.

Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor with Thermo Scientific™ Nunc™ PETG Roller Bottles. Virtually unbreakable PETG provides added assurance that valuable contents are safe during laboratory and industrial-scale applications including vaccines, cell culture expansion and production of other biologics.

Key Features:

  • Molded of durable PETG
  • Can be frozen down to -40°C — supports freeze thaw cell release methods, reducing reliance on trypsin
  • Provides solutions for all scales with wide range of surface areas from 1050 cm² to 4200 cm²
  • Reduces wrist strain and increases productivity with quick-action ergonomic closure
  • Can be traced by lot number on every bottle

Recommended for:

  • Cell culture expansion
  • Vaccine production
  • Antibody production

Warranty: 90 days

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Bottle SurfacePleated
Capacity (English)169.07 fl. oz.
Cell Growth Area4200 cm²
Closure TypeQuick-Action Ergonomic
Description4260-22
DisposableSingle-use
No. per Case22/Cs.
No. per Pack22 Pk.
Packaging22/Cs.
Product TypeRoller Bottle
ShapeRound
Capacity (Metric)5 L
Closure IncludedYes
ColorClear
GraduatedYes
MaterialPETG
MouthWide
SterilitySterile
Unit SizeCase of 22