This product ships via ground from US northeast.
hydrogen peroxide, 3 wt.% solution in water, stabilized
hydrogen peroxide, 3 wt.% solution in water, stabilized
hydrogen peroxide, 3 wt.% solution in water, stabilized
Thermo Scientific Chemicals

hydrogen peroxide, 3 wt.% solution in water, stabilized

CAS: 7722-84-1 | H2O2 | 34.014 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
1 L
Packaging:
Plastic bottle
Catalog number 426000010
Price (USD)
110.65
Special offer
Online exclusive
Ends: 28-Dec-2026
130.00
Save 19.35
Each
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
110.65
Special offer
Online exclusive
Ends: 28-Dec-2026
130.00
Save 19.35
Each
Chemical Identifiers
CAS7722-84-1
IUPAC Nameperoxol
Molecular FormulaH2O2
InChI KeyMHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILESOO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Solution
StabilizerApprox. 300 ppm
Titration with KMnO43 to 4 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only