Dimethyl chlorophosphate, 96%
Dimethyl chlorophosphate, 96%
Dimethyl chlorophosphate, 96%
Thermo Scientific Chemicals

Dimethyl chlorophosphate, 96%

CAS: 813-77-4 | C2H6ClO3P | 144.49 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 426140250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS813-77-4
IUPAC Namedimethyl phosphorochloridate
Molecular FormulaC2H6ClO3P
InChI KeyNGFFLHMFSINFGB-UHFFFAOYSA-N
SMILESCOP(Cl)(=O)OC
さらに表示
Appearance (Color)Clear colorless to yellow-brown
Infrared spectrumConforms
Refractive index1.4120 to 1.4140 (20°C, 589 nm)
Titration Argentometric>=95.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only