Methyl 2-amino-5-chlorobenzoate, 95%, Thermo Scientific™
Methyl 2-amino-5-chlorobenzoate, 95%, Thermo Scientific™
Methyl 2-amino-5-chlorobenzoate, 95%, Thermo Scientific™
Acros Organics

Methyl 2-amino-5-chlorobenzoate, 95%, Thermo Scientific™

 
Have Questions?
Catalog NumberQuantityPackaging
426160010
also known as 42616
1 gGlass bottle
Catalog number 426160010
also known as 42616
HSN 29224990
Price (INR)
15,525.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
15,525.00
Each
Chemical Identifiers
CAS5202-89-1
IUPAC Namemethyl 2-amino-5-chlorobenzoate
Molecular FormulaC8H8ClNO2
InChI KeyIGHVUURTQGBABT-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(Cl)=CC=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to beige to light brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
NMR (1H-NMR)Consistent with structure
GC>=94 %
View more
 
RUO – Research Use Only