1,4-Pentadien-3-ol, 97%, stabilized
1,4-Pentadien-3-ol, 97%, stabilized
1,4-Pentadien-3-ol, 97%, stabilized
Thermo Scientific Chemicals

1,4-Pentadien-3-ol, 97%, stabilized

CAS: 922-65-6 | C5H8O | 84.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 426240050
Price (USD)
119.65
Online exclusive
133.00
Save 13.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
119.65
Online exclusive
133.00
Save 13.35
Each
Chemical Identifiers
CAS922-65-6
IUPAC Namepenta-1,4-dien-3-ol
Molecular FormulaC5H8O
InChI KeyICMWSAALRSINTC-UHFFFAOYSA-N
SMILESOC(C=C)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless to yellow
Refractive index1.4440 to 1.4460 (20°C, 589 nm)
Stabilizer0.2 to 0.8 % Hydroquinone
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only