Thermo Scientific Chemicals

Methyl (R)-(+)-3-BOC-2,2-dimethyl-4-oxazolidinecarboxylate, 95%, Thermo Scientific Chemicals

製品番号(カタログ番号): 426350050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl (R)-(+)-3-BOC-2,2-dimethyl-4-oxazolidinecarboxylate, 95%, Thermo Scientific Chemicals

製品番号(カタログ番号): 426350050
5 g, Glass bottle, Each
数量
パッケージ
製品番号(カタログ番号): 426350050

化学物質識別子

CAS
95715-86-9
IUPAC Name
3-tert-butyl 4-methyl 2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate
Molecular Formula
C12H21NO5
InChI Key
ZNBUXTFASGDVCL-UHFFFAOYNA-N
SMILES
COC(=O)C1COC(C)(C)N1C(=O)OC(C)(C)C
Appearance (Color)
Clear colorless to yellow
Appearance (Form)
Liquid
Infrared spectrum
Conforms
NMR (1H-NMR)
Consistent with structure
Specific optical rotation
+53° to +57° (20°C, 589 nm) (C=1, Chloroform)

概要

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text