3-(Trimethoxysilyl)propyl acrylate, 90%, stabilized
3-(Trimethoxysilyl)propyl acrylate, 90%, stabilized
3-(Trimethoxysilyl)propyl acrylate, 90%, stabilized
Thermo Scientific Chemicals

3-(Trimethoxysilyl)propyl acrylate, 90%, stabilized

CAS: 4369-14-6 | C9H18O5Si | 234.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 426600050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4369-14-6
IUPAC Name3-(trimethoxysilyl)propyl prop-2-enoate
Molecular FormulaC9H18O5Si
InChI KeyKBQVDAIIQCXKPI-UHFFFAOYSA-N
SMILESCO[Si](CCCOC(=O)C=C)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4280 to 1.4300 (20°C, 589 nm)
StabilizerStabilized with BHT
GC>=90 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only