Di-tert-butyl dicarbonate, 1M solution in THF, AcroSeal™
Di-tert-butyl dicarbonate, 1M solution in THF, AcroSeal™
Di-tert-butyl dicarbonate, 1M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Di-tert-butyl dicarbonate, 1M solution in THF, AcroSeal™

CAS: 24424-99-5 | C10H18O5 | 218.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 426801000
Price (USD)
78.65
Online exclusive
87.10
Save 8.45
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
78.65
Online exclusive
87.10
Save 8.45
Each
Chemical Identifiers
CAS24424-99-5
IUPAC Namedi-tert-butyl dicarbonate
Molecular FormulaC10H18O5
InChI KeyDYHSDKLCOJIUFX-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)OC(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Titration by coupling0.90 to 1.10 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only