tert-Butylsulfinamide, 98%
tert-Butylsulfinamide, 98%
tert-Butylsulfinamide, 98%
Thermo Scientific Chemicals

tert-Butylsulfinamide, 98%

CAS: 146374-27-8 | C4H11NOS | 121.20 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 426830250
Price (JPY)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS146374-27-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to tan
Appearance (Form)Powder or crystals
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only