Ethyl 2-oximinooxamate, 98%, Thermo Scientific Chemicals
Ethyl 2-oximinooxamate, 98%, Thermo Scientific Chemicals
Ethyl 2-oximinooxamate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 2-oximinooxamate, 98%, Thermo Scientific Chemicals

CAS: 10489-74-4 | Molecular Formula: C4H8N2O3 | Molecular Weight: 132.12 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4268800505 gGlass bottle
Catalog number 426880050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10489-74-4
IUPAC Nameethyl (N'-hydroxycarbamimidoyl)formate
Molecular FormulaC4H8N2O3
InChI KeyQGYKRMZPOOILBA-UHFFFAOYSA-N
SMILESCCOC(=O)C(N)=NO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point96°C to 103°C
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Appearance (Color)White to yellow

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only