Di-tert-butyl N,N-diisopropylphosphoramidite, 95%
Di-tert-butyl N,N-diisopropylphosphoramidite, 95%
Di-tert-butyl N,N-diisopropylphosphoramidite, 95%
Thermo Scientific Chemicals

Di-tert-butyl N,N-diisopropylphosphoramidite, 95%

CAS: 137348-86-8 | C14H32NO2P | 277.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 426890050
Price (USD)
124.65
Online exclusive
138.00
Save 13.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
124.65
Online exclusive
138.00
Save 13.35
Each
Chemical Identifiers
CAS137348-86-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4390 to 1.4410 (20°C, 589 nm)
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only