4-Methoxy-2-methylbenzaldehyde, 97%, Thermo Scientific Chemicals
4-Methoxy-2-methylbenzaldehyde, 97%, Thermo Scientific Chemicals
4-Methoxy-2-methylbenzaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Methoxy-2-methylbenzaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantityPackaging
4270200505 gGlass bottle
Catalog number 427020050
Price (EUR)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS52289-54-0
IUPAC Name4-methoxy-2-methylbenzaldehyde
Molecular FormulaC9H10O2
InChI KeyWICYVKGMEJSDAO-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
GC>=96.0 %
Appearance (Form)Liquid
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only