Boron trifluoride etherate, ca. 48% BF3, AcroSeal™
Boron trifluoride etherate, ca. 48% BF3, AcroSeal™
Boron trifluoride etherate, ca. 48% BF3, AcroSeal™
Thermo Scientific Chemicals

Boron trifluoride etherate, ca. 48% BF3, AcroSeal™

CAS: 109-63-7 | C4H10BF3O | 141.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 427118000
Price (USD)
179.00
Each
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
179.00
Each
Chemical Identifiers
CAS109-63-7
IUPAC Nameethoxyethane; trifluoroborane
Molecular FormulaC4H10BF3O
InChI KeyKZMGYPLQYOPHEL-UHFFFAOYSA-N
SMILESFB(F)F.CCOCC
View more
SpecificationsSpecification SheetSpecification Sheet
Assay47.5 to 49 % (BF3)
Appearance (Form)Liquid
Appearance (Color)Clear colorless to straw yellow
Refractive index1.3429 to 1.3449 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only