Benzylamine, 99.5+%, AcroSeal™
Benzylamine, 99.5+%, AcroSeal™
Benzylamine, 99.5+%, AcroSeal™
Thermo Scientific Chemicals

Benzylamine, 99.5+%, AcroSeal™

CAS: 100-46-9 | C7H9N | 107.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 427238000
Price (USD)
-
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS100-46-9
IUPAC Name1-phenylmethanamine
Molecular FormulaC7H9N
InChI KeyWGQKYBSKWIADBV-UHFFFAOYSA-N
SMILESNCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.3 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.5424 to 1.5444 (20°C, 589 nm)
Infrared spectrumConforms
GC>=99.50 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only