trans-N,N'-Dimethyl-1,2-cyclohexanediamine
trans-N,N'-Dimethyl-1,2-cyclohexanediamine
trans-N,N'-Dimethyl-1,2-cyclohexanediamine
Thermo Scientific Chemicals

trans-N,N'-Dimethyl-1,2-cyclohexanediamine

CAS: 67579-81-1 | C8H18N2 | 142.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 427310050
Price (USD)
183.65
Special offer
Online exclusive
Ends: 28-Dec-2026
216.00
Save 32.35
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
183.65
Special offer
Online exclusive
Ends: 28-Dec-2026
216.00
Save 32.35
Each
Chemical Identifiers
CAS67579-81-1
IUPAC NameN1,N2-dimethylcyclohexane-1,2-diamine
Molecular FormulaC8H18N2
InChI KeyJRHPOFJADXHYBR-UHFFFAOYNA-N
SMILESCNC1CCCCC1NC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4710 to 1.4730 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only