Tetramethylsilane, 99%, AcroSeal™
Tetramethylsilane, 99%, AcroSeal™
Tetramethylsilane, 99%, AcroSeal™
Thermo Scientific Chemicals

Tetramethylsilane, 99%, AcroSeal™

CAS: 75-76-3 | C4H12Si | 88.23 g/mol
Have Questions?
Catalog NumberQuantityPackaging
427331000100 mLAcroSeal™ Glass Bottle
Catalog number 427331000
Price (USD)
88.65
Special offer
Online exclusive
Ends: 28-Dec-2026
104.00
Save 15.35
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
88.65
Special offer
Online exclusive
Ends: 28-Dec-2026
104.00
Save 15.35
Each
Chemical Identifiers
CAS75-76-3
IUPAC Nametetramethylsilane
Molecular FormulaC4H12Si
InChI KeyCZDYPVPMEAXLPK-UHFFFAOYSA-N
SMILESC[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.3575 to 1.3595 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only