3-Aminophenylacetic acid, 97%
3-Aminophenylacetic acid, 97%
3-Aminophenylacetic acid, 97%
Thermo Scientific Chemicals

3-Aminophenylacetic acid, 97%

CAS: 14338-36-4 | C8H9NO2 | 151.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass Bottle
Catalog number 427610010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS14338-36-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to brown
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
Melting point147°C to 154°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only