3-Aminophenylacetic acid, 97%
3-Aminophenylacetic acid, 97%
3-Aminophenylacetic acid, 97%
Thermo Scientific Chemicals

3-Aminophenylacetic acid, 97%

CAS: 14338-36-4 | C8H9NO2 | 151.17 g/mol
数量:
1 g
10 g
パッケージ:
Glass Bottle
製品番号(カタログ番号) 427610100
価格(JPY)
-
数量:
10 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS14338-36-4
Appearance (Color)Off-white to brown
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
Melting point147°C to 154°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only