3-Bromo-2-methylpropene, 97%
3-Bromo-2-methylpropene, 97%
3-Bromo-2-methylpropene, 97%
Thermo Scientific Chemicals

3-Bromo-2-methylpropene, 97%

CAS: 1458-98-6 | C4H7Br | 135.00 g/mol
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 427650250
Price (JPY)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1458-98-6
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4710 to 1.4730 (20°C, 589 nm)
GC>=96.0 %
StabilizerApprox 500 ppm hydroquinone
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only