Isopropenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Isopropenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Isopropenylmagnesium bromide, 0.5M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Isopropenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

CAS: 13291-18-4 | C3H5BrMg | 145.28 g/mol
数量:
100 mL
800 mL
パッケージ:
すべて表示
AcroSeal™ Glass bottle
製品番号(カタログ番号) 427751000
価格(JPY)
-
数量:
100 mL
パッケージ:
AcroSeal™ Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13291-18-4
IUPAC Namebromo(prop-1-en-2-yl)magnesium
Molecular FormulaC3H5BrMg
InChI KeyLFTYALBVGVNGLI-UHFFFAOYSA-M
SMILESCC(=C)[Mg]Br
さらに表示
Appearance (Color)Clear light yellow to orange to grey to brown
Appearance (Form)Solution
Titration Active base0.45 to 0.60 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only