Hexylmagnesium bromide, 0.8M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Hexylmagnesium bromide, 0.8M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Hexylmagnesium bromide, 0.8M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hexylmagnesium bromide, 0.8M solution in THF, AcroSeal™, Thermo Scientific Chemicals

CAS: 3761-92-0 | Molecular Formula: C6H13BrMg | Molecular Weight: 189.38 g/mol
Have Questions?
Catalog NumberQuantityPackaging
427761000100 mLAcroSeal™ Glass bottle
Catalog number 427761000
Price (USD)
65.65
Online exclusive
72.60
Save 6.95
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
65.65
Online exclusive
72.60
Save 6.95
Each
Chemical Identifiers
CAS3761-92-0
IUPAC Namebromo(hexyl)magnesium
Molecular FormulaC6H13BrMg
InChI KeySFXUWNMDTWQSHE-UHFFFAOYSA-M
SMILESCCCCCC[Mg]Br
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Grignard0.6 to 1 M
Appearance (Color)Brown
Appearance (Form)Solution

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only