3-Fluorophenylmagnesium bromide, 1M solution in THF, AcroSeal™
3-Fluorophenylmagnesium bromide, 1M solution in THF, AcroSeal™
3-Fluorophenylmagnesium bromide, 1M solution in THF, AcroSeal™
Thermo Scientific Chemicals

3-Fluorophenylmagnesium bromide, 1M solution in THF, AcroSeal™

CAS: 17318-03-5 | C6H4BrFMg | 199.31 g/mol
Have Questions?
Catalog NumberQuantity
427821000100 mL
Catalog number 427821000
Price (EUR)
165,00
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
165,00
Each
Chemical Identifiers
CAS17318-03-5
IUPAC Namebromo(3-fluorophenyl)magnesium
Molecular FormulaC6H4BrFMg
InChI KeyVFPDAAQAKDGSHQ-UHFFFAOYSA-M
SMILESFC1=CC([Mg]Br)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Grignard1 M Exact strength on the certificate of analysis
Appearance (Color)Yellow to brown
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only