7-Hydroxycoumarinyl-4-acetic acid, 97%
7-Hydroxycoumarinyl-4-acetic acid, 97%
7-Hydroxycoumarinyl-4-acetic acid, 97%
Thermo Scientific Chemicals

7-Hydroxycoumarinyl-4-acetic acid, 97%

CAS: 6950-82-9 | C11H8O5 | 220.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 427900010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6950-82-9
IUPAC Name2-(7-hydroxy-2-oxo-2H-chromen-4-yl)acetic acid
Molecular FormulaC11H8O5
InChI KeyBNHPMQBVNXMPDU-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC(=O)OC2=CC(O)=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=96.0 %
Appearance (Color)Off-white to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only