Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™
Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™
Thermo Scientific Chemicals

Zirconium(IV) propoxide, ca. 70%, solution in 1-Propanol, AcroSeal™

CAS: 23519-77-9 | C12H28O4Zr | 327.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 427938000
Price (EUR)
194,00
Each
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (EUR)
194,00
Each
Chemical Identifiers
CAS23519-77-9
IUPAC Nametetrapropoxyzirconium
Molecular FormulaC12H28O4Zr
InChI KeyXPGAWFIWCWKDDL-UHFFFAOYSA-N
SMILESCCCO[Zr](OCCC)(OCCC)OCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Assay27.0 to 29.0 % (ZrO2)
Appearance (Color)Colorless to yellow to amber
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only