Ethyl trans-beta-methylcinnamate, 97%
Ethyl trans-beta-methylcinnamate, 97%
Ethyl trans-beta-methylcinnamate, 97%
Thermo Scientific Chemicals

Ethyl trans-beta-methylcinnamate, 97%

CAS: 1504-72-9 | C12H14O2 | 190.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 428000010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1504-72-9
IUPAC Nameethyl 3-phenylbut-2-enoate
Molecular FormulaC12H14O2
InChI KeyBSXHSWOMMFBMLL-UHFFFAOYSA-N
SMILESCCOC(=O)C=C(C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=96.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only