Thermo Scientific™
Edge-Rite™ Disposable Microtome Blades
Related applications:
Anatomical Pathology
Experience consistent quality from blade to blade and sample to sample with Thermo Scientific™ Edge-Rite™ Disposable Microtome Blades.
Back to top
  • Surgical-grade steel
  • High- and low-profile blades are specially coated to increase longevity and optimize section quality
Low profile blades:
  • Blades are 20% thicker than traditional blades to increase knife edge stability, which allows for excellent ribboning and facilitates usage in cryostats
  • Blades are 5% longer, allowing for more sections per blade
  • Blades fit any low-profile blade holder