Ammonia, ca. 7N solution in methanol, AcroSeal™
Ammonia, ca. 7N solution in methanol, AcroSeal™
Ammonia, ca. 7N solution in methanol, AcroSeal™
Thermo Scientific Chemicals

Ammonia, ca. 7N solution in methanol, AcroSeal™

CAS: 7664-41-7 | H3N | 17.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Catalog number 428388000
Price (EUR)
162,65
Online exclusive
180,00
Save 17,35 (10%)
Each
Quantity:
800 mL
Request bulk or custom format
Price (EUR)
162,65
Online exclusive
180,00
Save 17,35 (10%)
Each
Chemical Identifiers
CAS7664-41-7
IUPAC Nameammonia
Molecular FormulaH3N
InChI KeyQGZKDVFQNNGYKY-UHFFFAOYSA-N
SMILESN
View more
SpecificationsSpecification SheetSpecification Sheet
Strength6.0 to 7.5 N
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only