Methyl cis-11-eicosenoate, 99%
Methyl cis-11-eicosenoate, 99%
Methyl cis-11-eicosenoate, 99%
Thermo Scientific Chemicals

Methyl cis-11-eicosenoate, 99%

CAS: 2390-09-2 | C21H40O2 | 324.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 428530010
Price (USD)
371.65
Special offer
Online exclusive
Ends: 28-Dec-2026
437.00
Save 65.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
371.65
Special offer
Online exclusive
Ends: 28-Dec-2026
437.00
Save 65.35
Each
Chemical Identifiers
CAS2390-09-2
IUPAC Namemethyl (11E)-icos-11-enoate
Molecular FormulaC21H40O2
InChI KeyRBKMRGOHCLRTLZ-ZHACJKMWSA-N
SMILESCCCCCCCC\C=C\CCCCCCCCCC(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless
Appearance (Form)Liquid
NMR (1H-NMR)Consistent with structure
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only