Methyl cis-11-eicosenoate, 99%
Methyl cis-11-eicosenoate, 99%
Methyl cis-11-eicosenoate, 99%
Thermo Scientific Chemicals

Methyl cis-11-eicosenoate, 99%

CAS: 2390-09-2 | C21H40O2 | 324.55 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 428530050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2390-09-2
IUPAC Namemethyl (11E)-icos-11-enoate
Molecular FormulaC21H40O2
InChI KeyRBKMRGOHCLRTLZ-ZHACJKMWSA-N
SMILESCCCCCCCC\C=C\CCCCCCCCCC(=O)OC
さらに表示
Infrared spectrumConforms
Appearance (Color)Clear colorless
Appearance (Form)Liquid
NMR (1H-NMR)Consistent with structure
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only