Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt, 95%
Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt, 95%
Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt, 95%
Thermo Scientific Chemicals

Ethylene glycol-bis(beta-aminoethyl ether)-N,N,N',N'-tetraacetic acid tetrasodium salt, 95%

CAS: 13368-13-3 | C14H20N2Na4O10 | 468.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 428570100
Price (USD)
93.65
Online exclusive
104.00
Save 10.35
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
93.65
Online exclusive
104.00
Save 10.35
Each
Chemical Identifiers
CAS13368-13-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
Loss on drying=<20 % (110°C, 16 hrs)
Assay18.46 to 20.82 % (ICP on dried sample)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only