Cyclopropylboronic acid pinacol ester, 95%
Cyclopropylboronic acid pinacol ester, 95%
Cyclopropylboronic acid pinacol ester, 95%
Thermo Scientific Chemicals

Cyclopropylboronic acid pinacol ester, 95%

CAS: 126689-01-8 | C9H17BO2 | 168.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 428590010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS126689-01-8
IUPAC Name2-cyclopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC9H17BO2
InChI KeyXGBMQBPLWXTEPM-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=94 %
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only