Triethyl borate, 97%, AcroSeal™, Thermo Scientific Chemicals
Triethyl borate, 97%, AcroSeal™, Thermo Scientific Chemicals
Triethyl borate, 97%, AcroSeal™, Thermo Scientific Chemicals
Triethyl borate, 97%, AcroSeal™, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethyl borate, 97%, AcroSeal™, Thermo Scientific Chemicals

CAS: 150-46-9 | Molecular Formula: C6H15BO3 | Molecular Weight: 145.99 g/mol
Have Questions?
Catalog NumberQuantityPackaging
428651000100 mLAcroSeal™ Glass Bottle
Catalog number 428651000
Price (USD)
46.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.10
Save 8.45
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
46.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.10
Save 8.45
Each
Chemical Identifiers
CAS150-46-9
IUPAC Nametriethyl borate
Molecular FormulaC6H15BO3
InChI KeyAJSTXXYNEIHPMD-UHFFFAOYSA-N
SMILESCCOB(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Refractive index1.3705 to 1.3775 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Color)Clear colorless

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only