Cyanogen bromide, 3M solution in dichloromethane, AcroSeal™
Cyanogen bromide, 3M solution in dichloromethane, AcroSeal™
Cyanogen bromide, 3M solution in dichloromethane, AcroSeal™
Thermo Scientific Chemicals

Cyanogen bromide, 3M solution in dichloromethane, AcroSeal™

CAS: 506-68-3 | BrCN | 105.93 g/mol
Have Questions?
Catalog NumberQuantity
428911000100 mL
Catalog number 428911000
Price (EUR)
62,65
Online exclusive
69,40
Save 6,75 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
62,65
Online exclusive
69,40
Save 6,75 (10%)
Each
Chemical Identifiers
CAS506-68-3
IUPAC Namecarbononitridic bromide
Molecular FormulaCBrN
InChI KeyATDGTVJJHBUTRL-UHFFFAOYSA-N
SMILESBrC#N
View more
SpecificationsSpecification SheetSpecification Sheet
Strength2.95 to 3.30 M
Appearance (Form)Solution
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only