Iodine monochloride, 1M solution in dichloromethane, AcroSeal™
Iodine monochloride, 1M solution in dichloromethane, AcroSeal™
Iodine monochloride, 1M solution in dichloromethane, AcroSeal™
Iodine monochloride, 1M solution in dichloromethane, AcroSeal™
Thermo Scientific Chemicals

Iodine monochloride, 1M solution in dichloromethane, AcroSeal™

CAS: 7790-99-0 | ClI | 162.35 g/mol
Have Questions?
Catalog NumberQuantityPackaging
428941000100 mLAcroSeal™ Glass Bottle
Catalog number 428941000
Price (USD)
73.65
Online exclusive
82.20
Save 8.55
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
73.65
Online exclusive
82.20
Save 8.55
Each
Chemical Identifiers
CAS7790-99-0
IUPAC Nameiodochlorane
Molecular FormulaClI
InChI KeyQZRGKCOWNLSUDK-UHFFFAOYSA-N
SMILESClI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Titration with Na2S2O30.95 to 1.05 M
Appearance (Color)Orange to red to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only