Diisopropylphosphine, 10 wt.% solution in hexanes, AcroSeal™
Diisopropylphosphine, 10 wt.% solution in hexanes, AcroSeal™
Diisopropylphosphine, 10 wt.% solution in hexanes, AcroSeal™
Thermo Scientific Chemicals

Diisopropylphosphine, 10 wt.% solution in hexanes, AcroSeal™

CAS: 20491-53-6 | C6H15P | 118.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
429041000100 mLAcroSeal™ Glass bottle
Catalog number 429041000
Price (USD)
-
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20491-53-6
IUPAC Namebis(propan-2-yl)phosphane
Molecular FormulaC6H15P
InChI KeyWDIIYWASEVHBBT-UHFFFAOYSA-N
SMILESCC(C)PC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Concentration9 to 11 wt%
NMR (31P-NMR)Consistent with structure
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only