Anisole, 99%, Extra Dry, AcroSeal™
Anisole, 99%, Extra Dry, AcroSeal™
Anisole, 99%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

Anisole, 99%, Extra Dry, AcroSeal™

CAS: 100-66-3 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Catalog number 429251000
Price (EUR)
47,65
Online exclusive
52,30
Save 4,65 (9%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
47,65
Online exclusive
52,30
Save 4,65 (9%)
Each
Chemical Identifiers
CAS100-66-3
IUPAC Nameanisole
Molecular FormulaC7H8O
InChI KeyRDOXTESZEPMUJZ-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
GC>=98.5 %
Appearance (Form)Liquid
Water=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only