Triethyl orthoformate, 98%, anhydrous, AcroSeal™
Triethyl orthoformate, 98%, anhydrous, AcroSeal™
Triethyl orthoformate, 98%, anhydrous, AcroSeal™
Triethyl orthoformate, 98%, anhydrous, AcroSeal™
Thermo Scientific Chemicals

Triethyl orthoformate, 98%, anhydrous, AcroSeal™

CAS: 122-51-0 | C7H16O3 | 148.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 429271000
Price (USD)
43.65
Special offer
Online exclusive
Ends: 28-Dec-2026
51.50
Save 7.85
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
43.65
Special offer
Online exclusive
Ends: 28-Dec-2026
51.50
Save 7.85
Each
Chemical Identifiers
CAS122-51-0
IUPAC Name(diethoxymethoxy)ethane
Molecular FormulaC7H16O3
InChI KeyGKASDNZWUGIAMG-UHFFFAOYSA-N
SMILESCCOC(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=97.5 %
Residue after evaporation=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only