Sodium ethoxide, 21% in ethanol, AcroSeal™
Sodium ethoxide, 21% in ethanol, AcroSeal™
Sodium ethoxide, 21% in ethanol, AcroSeal™
Sodium ethoxide, 21% in ethanol, AcroSeal™
Thermo Scientific Chemicals

Sodium ethoxide, 21% in ethanol, AcroSeal™

CAS: 141-52-6 ∣ C2H5NaO ∣ 68.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Catalog number 429301000
Price (USD)
43.80
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
43.80
Each
Chemical Identifiers
CAS141-52-6
IUPAC Namesodium ethanolate
Molecular FormulaC2H5NaO
InChI KeyQDRKDTQENPPHOJ-UHFFFAOYSA-N
SMILES[Na+].CC[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay darken on storage
Titration with HCl20.0 to 22.0 %
NaOH + Na2CO3 (as NaOH)=<0.5 %
Appearance (Color)Yellow to brown
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use