4-Methyl-1,3-thiazole-2-carboxylic acid, 97%
4-Methyl-1,3-thiazole-2-carboxylic acid, 97%
4-Methyl-1,3-thiazole-2-carboxylic acid, 97%
Thermo Scientific Chemicals

4-Methyl-1,3-thiazole-2-carboxylic acid, 97%

CAS: 14542-16-6 | C5H5NO2S | 143.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 429360050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14542-16-6
IUPAC Name4-methyl-1,3-thiazole-2-carboxylic acid
Molecular FormulaC5H5NO2S
InChI KeyGNGDWDFLILPTKL-UHFFFAOYSA-N
SMILESCC1=CSC(=N1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=96.0 %
Appearance (Color)White to off-white to light yellow
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only