4-Methylpyridine-3-carboxylic acid, 97%
4-Methylpyridine-3-carboxylic acid, 97%
4-Methylpyridine-3-carboxylic acid, 97%
Thermo Scientific Chemicals

4-Methylpyridine-3-carboxylic acid, 97%

CAS: 3222-50-2 | C7H7NO2 | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 429420010
Price (USD)
213.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
213.00
Each
Chemical Identifiers
CAS3222-50-2
SpecificationsSpecification SheetSpecification Sheet
Melting point212°C to 220°C
Appearance (Color)White to tan
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only