Tetrahydropyran-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Tetrahydropyran-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrahydropyran-4-sulfonyl chloride, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 429440050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Nameoxane-4-sulfonyl chloride
Molecular FormulaC5H9ClO3S
InChI KeyQQSBPTQNEOJFBO-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1CCOCC1
Molecular Weight (g/mol)184.63
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Colorless to light brown
GC>=96.0 %
Appearance (Form)Low melting solid
Melting point35°C to 41°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only