6-Bromoisoquinoline, 97%
6-Bromoisoquinoline, 97%
6-Bromoisoquinoline, 97%
Thermo Scientific Chemicals

6-Bromoisoquinoline, 97%

CAS: 34784-05-9 | C9H6BrN | 208.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 429460050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS34784-05-9
IUPAC Name6-bromoisoquinoline
Molecular FormulaC9H6BrN
InChI KeyZTEATMVVGQUULZ-UHFFFAOYSA-N
SMILESBrC1=CC=C2C=NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting solid
HPLC>=96.0 %
Infrared spectrumConforms
Melting point40°C to 45°C
Appearance (Color)Orange to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only