1H-Benzimidazole-4-carboxylic acid, 97%
1H-Benzimidazole-4-carboxylic acid, 97%
1H-Benzimidazole-4-carboxylic acid, 97%
Acros Organics

1H-Benzimidazole-4-carboxylic acid, 97%

CAS: 46006-36-4 | C8H6N2O2 | 162.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 429510010
HSN 29339900
Price (INR)
62,282.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
62,282.00
Each
Specifications
Chemical Name or Material1H-Benzimidazole-4-carboxylic acid
CAS46006-36-4
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only