2-Pirimidinacarbonitrilo, 98 %, Thermo Scientific Chemicals
2-Pirimidinacarbonitrilo, 98 %, Thermo Scientific Chemicals
2-Pirimidinacarbonitrilo, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Pirimidinacarbonitrilo, 98 %, Thermo Scientific Chemicals

CAS: 14080-23-0 | C5H3N3 | 105.10 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
25 g
Envase:
Frasco de vidrio
Número de catálogo 429820250
Precio (USD)
-
Cantidad:
25 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS14080-23-0
IUPAC Namepyrimidine-2-carbonitrile
Molecular FormulaC5H3N3
InChI KeyIIHQNAXFIODVDU-UHFFFAOYSA-N
SMILESN#CC1=NC=CC=N1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder or low melting solid
Infrared spectrumConforms
Melting point35°C to 42°C
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only