6-Bromo-2-pyridinecarbonitrile, 98%
6-Bromo-2-pyridinecarbonitrile, 98%
6-Bromo-2-pyridinecarbonitrile, 98%
Thermo Scientific Chemicals

6-Bromo-2-pyridinecarbonitrile, 98%

CAS: 122918-25-6 | C6H3BrN2 | 183.01 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4299900101 gGlass bottle
Catalog number 429990010
Price (USD)
55.65
Special offer
Online exclusive
Ends: 28-Dec-2026
65.40
Save 9.75
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
55.65
Special offer
Online exclusive
Ends: 28-Dec-2026
65.40
Save 9.75
Each
Chemical Identifiers
CAS122918-25-6
IUPAC Name6-bromopyridine-2-carbonitrile
Molecular FormulaC6H3BrN2
InChI KeyHNEBPTAKURBYRM-UHFFFAOYSA-N
SMILESBrC1=CC=CC(=N1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point98°C to 103°C
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only