1H-Indazole-6-carboxylic acid, 97%
1H-Indazole-6-carboxylic acid, 97%
1H-Indazole-6-carboxylic acid, 97%
Thermo Scientific Chemicals

1H-Indazole-6-carboxylic acid, 97%

CAS: 704-91-6 | C8H6N2O2 | 162.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 430120010
Price (EUR)
64,50
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
64,50
Each
Chemical Identifiers
CAS704-91-6
IUPAC Name1H-indazole-6-carboxylic acid
Molecular FormulaC8H6N2O2
InChI KeyDNCVTVVLMRHJCJ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C2C=NNC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only